Print…
Redox/ORP/temperature electrodes, Orion™
Fornecedor: Thermo Orion
9179BN 9179BNMD 9180BNMD
ORIO9179BNEA 455 EUR
ORIO9179BN 662-1456 662-1457
Redox/ORP/temperature electrodes, Orion™
Eléctrodos Redox Electrodes
These ORP electrodes combine a platinum redox sensing electrode and a silver/silver chloride reference electrode in one body.
  • Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • ORP Triode electrodes have a built-in thermistor for measuring temperature
The 9180BNMD and 9179BNMD ORP/ATC Triode electrodes have a temperature connector that is compatible with the Star Series meters.
The 9180BN and 9179BN ORP/Mais informação do produto