Print…
Combined ion selective electrodes, 500 series
Fornecedor: WTW
106395
662-0381EA 1270 EUR
662-0381
Combined ion selective electrodes, 500 series
Eléctrodos Eléctrodos ISE
These 500 series half cells require a separate reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated reference electrode).
  • Ideal for challenging ISE applications
Informações de encomenda: Reference electrodes, ionic strength adapters and standard solutions are not included. Please contact Avantor for further information.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable