Print…
Células de fluxo contínuo para medições de absorção
Fornecedor: HELLMA
170700-0.1-40 170700-2-40 170000-2-40 170000-1-40 1767001550-40 170700-0.5-40 178710-1085-40 137-1-40 137-2-40 137-5-40 137-10-40 175-10-15-40 175-10-85-40 176-50-85-40 1767005-15-40 178712-1085-40 170700-0.2-40 170-700-1-40 176-10-15-40 178710-1015-40 1767008510-40 1767001510-40 176-50-15-40 176700-5-85-40 176-760-85-40 176-761-15-40 178712-10-20 170700-0.2-K-4 170700-0.5-K-4 1767001015-K-4 1767001085-K-4 1767005-15-K-4 178710-1015K-4
634-9823EA 1250 EUR
634-9823 634-9827 634-9821 634-9820 634-9936 634-9825 634-0869 634-0859 634-0982 634-0983 634-0830 634-0860 634-0861 634-0863 634-9103 634-0989 634-9824 634-9826 634-9107 634-9945 634-9102 634-9104 634-9108 634-9100 634-0984 634-0985 HELL178712-10-20 HELL170700-0.2-K-4 HELL170700-0.5-K-4 HELL1767001015-K-4 HELL1767001085-K-4 HELL1767005-15-K-4 HELL178710-1015K-4
Células de fluxo contínuo para medições de absorção
Cuvetes Cuvetes para espetroscopia
Hellma® high precision quartz glass flow-through cells for absorption measurements in the UV/Vis range and/or fluorescence measurements, 0,1 to 50 mm path length, with inlet and outlet nozzle, or 2 screw connections and FEP tubes.
  • Cell made of high performance quartz glass
  • Wavelength range from 200 to 2500 nm
  • 0,1 to 50 mm path length
  • With inlet and outlet nozzle, or 2 screw connections and FEP tubes
  • Volume from 6,2 to 2600 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.