Print…
VWR®, Cuvetes descartáveis
Fornecedor: VWR Collection
634-0678EA 16.9 EUR
634-0678 634-0675 634-0676 634-0677
VWR®, Cuvetes descartáveis
Cuvetes Cuvetes para espetroscopia
Disposable cuvettes in either optical polystyrene (PS) or polymethyl methacrylate (PMMA), for use with most polar solvents, acids and alkaline solutions.
  • Minimum variation of extinction values
  • Recessed windows, clear and clean optical path
  • Optimum transmittance and reduced risk of contamination
For determinations in water analysis, chemistry and life science applications.
Embalagem: Resealable – cuvettes are assorted by cavity numbers.