Print…
Tiras indicadoras de temperatura
Fornecedor: Testo
0646.0072 0646.0073 0646.0074 0646.0075 0646.0076 0646.0077
620-9122EA 36 EUR
620-9122 620-9123 620-9124 620-9125 620-9126 620-9127
Tiras indicadoras de temperatura
Tiras indicadoras de temperatura
These indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are particularly suitable for monitoring temperature in small objects.
  • Change in colour within two seconds
  • Practical booklet with 10 indicators
  • Maximum storage temperature: 25 °C; storage in a refrigerator is recommended