Print…
Termómetro compacto
Fornecedor: Testo
0900.0530
620-3123EA 53 EUR
620-3123
Termómetro compacto
Termómetros Termómetros Digitais
Small and handy mini thermometer with min./max. alarm.
  • Adjustable min./max. alarm: Acoustic signal sounds when the temperature exceeds or falls below the individually adjustable limit values
  • Permanently connected probe (penetration probe with 80 cm cable)
  • Probe and cable can be stored easily on the temperature measuring instrument
  • With clip for set-up, wall mounting or attachment
The mini alarm thermometer is suitable for temperature measurement in air, in soft powdery substances and in liquids. Using the clips supplied, the mini alarm thermometer canMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Temperatura de serviço –10...+50 °C
Sonda L×Ø: 190×3 mm
Probe cable length 80 cm
Ecrã LCD
Battery type 2 batteries type micro AAA 1,5 V (LR03)
Battery life 500 hours
Housing material ABS
W×D×H 64×18×100 mm
Peso 135 g