Print…
Reading pocket magnifiers, mobilux
Fornecedor: ESCHENBACH OPTIK
15117 15113 151112 15112 15116 15114 15122
630-1313EA 86.1 EUR
630-1313 630-1312 630-2178 630-2179 630-2180 620-2616 ESCH15122
Reading pocket magnifiers, mobilux
Lupas
LED illumination and coated lens on both sides with Ceratec® anti-scratch coating for optimised performances.
  • Exchangeable white light LED
  • Attachable yellow filter of 50000 hours
  • Easy handling and large light switch
Fornecimento: Supplied with two AA batteries and case.