Print…
Cryoscopes, CryoStar
Fornecedor: GERBER FUNKE
7150
614-2060EA 4530 EUR
614-2060
Cryoscopes, CryoStar
Crioscópios
Quick and reliable measurement of the freezing point of milk. The cooling system provides quick operational readiness even at high ambient temperatures (up to 32 °C).
  • Easy to use, uncomplicated operation
  • Multi-language menu
  • Automatic calibration and settings history saved to non volatile memory
  • Software included for mapping the freezing curve on a PC
Certificações: Reference measurement in accordance with DIN/ISO/IDF5764
Encomendar
ESPECIFICAÇÕES
Volume da amostra 2,0 - 2,5 ml
Environmentally Preferable
Tempo de refrigeração 15 min
Capacidade para amostras Up to 40 samples/h, typically 30/h
Unidades °C, % infitration water, date, time
Resolução 0,0001 °C
Gama −1,5000...0,0000 °C
Repetibilidade ±0,002 °C
Ecrã Graphic colour display
L×P×A 290×380×190 mm
Peso 12 kg