Print…
Milk butyrometer
Fornecedor: GERBER FUNKE
3155-C-O 3151 3152-O 3156 3153 3154 3151-C 3155-C 3154-O 3158 3157 3155 3152 3151-C-O 3157-C 3157-C-O
614-2006EA 30.6 EUR
614-2006 614-0089 709-0142 614-0094 614-0091 614-0092 GERB3151-C GERB3155-C 709-0143 614-0096 614-0095 614-0093 614-0090 GERB3151-C-O GERB3157-C GERB3157-C-O
Milk butyrometer
Butirómetros
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).