Print…
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Fornecedor: EPPENDORF
0030078.691 0030078.705 0030078.713 0030078.721 0030078.730 0030078.748
613-6791EA 207 EUR
613-6791 613-6792 613-6793 613-6794 613-6795 613-6796
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Pontas de pipetas
Eppendorf ep Dualfilter T.I.P.S.® SealMax® Racks provide advanced self-sealing pipette tips in 'PCR clean and Sterile' grade, featuring unique two-phase filters for unparalleled protection against contamination in sensitive lab applications.
  • Reliable protection from liquids, aerosols, and biomolecules
  • No PCR inhibiting additives
  • Protection of pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity