Print…
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
Fornecedor: Thermo Fisher Scientific
4672010BT 4672020BT 4672030BT 4672040BT 4672050BT 4672060BT 4672070BT 4672080BT 4672090BT 4672100BT
613-5767EA 1903 EUR
613-5767 613-5770 613-5768 613-5771 613-5764 613-5769 613-5772 613-5765 613-5763 613-5766
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
Pipetas Micropipetas
These pipettes are ideal for quickly and efficiently performing sample transfers between virtually any tube, rack, microplate or horizontal gel box. Adjustable tip spacing allows the user to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • ClipTip Interlocking Technology: Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every
Mais informação do produto