Print…
Buretas, torneira lateral, Schellbach, BLAUBRAND®
Fornecedor: Brand
13844 13847 13848
612-9941EA 135 EUR
612-9941 612-9942 612-9943
Buretas, torneira lateral, Schellbach, BLAUBRAND®
Buretas
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represents the optimum combination of resistance and readability
Length: 800 mm.
Comprimento: 800 mm
Certificações: DIN EN ISO 385
Informações de encomenda: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.