Print…
VWR®, Transfer Pipettes, Graduated
Fornecedor: VWR Collection
612-6803EA 27.1 EUR
612-6803 612-6805 612-6804 612-6806
VWR®, Transfer Pipettes, Graduated
Pipetas Pipetas Pasteur
These LDPE disposable, unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Non toxic and inert to biological fluids and most acids
  • Can be sealed, refrigerated and gas sterilised
  • No bulb to insert or remove
  • Uniform drop size
They work well whenever there is a need for quick, safe transfer of fluids.
Certificações: Compliance to US FDA 21 CFR.