Print…
Pipette controller, S1
Fornecedor: Thermo Fisher Scientific
9501 9511 9521 9531 9541
612-3983EA 400 EUR
612-3983 612-3984 612-3985 612-3986 612-3987
Pipette controller, S1
Pipetas Controladores de Pipetas
The S1 pipette controller combines pipetting comfort and high performance. For use with graduated and volumetric glass and plastic serological pipettes from 1 to 100 ml.
  • Lightweight ergonomic design; large illuminated LCD display
  • 8 speed alternatives for both aspirating and dispensing allowing total control of pipetting
  • Customisable ID area, five colour options permit easy recognition of ownership and/or task designation
  • Autoclavable silicone adapter
  • Heavy duty lithium-ion battery with fast recharging (3 hours) and 15 hours of continuous
Mais informação do produto