Print…
Burettes, straight stopcock
Fornecedor: HIRSCHMANN
3100160 3100170 3100175 3100260 3100270 3100275
612-3526EA 62.8 EUR
612-3526 612-3527 612-9810 612-3529 612-3531 612-3532
Burettes, straight stopcock
Buretas
DURAN®, class B.
  • Calibrated to deliver 'Ex'
  • Short marks at major graduations, black graduations
Scale length : 500 mm.
Certificações: DIN EN ISO 385.