Print…
Pipetas volumétricas, BLAUBRAND®, 2 marcas, classe AS
Fornecedor: Brand
29721 29722 29723 29725 29727 29732 29733 29734 29735 29738
612-2726EA 11.2 EUR
612-2726 612-2727 612-2728 612-2729 612-2730 612-2731 BRAA29733 612-2732 612-2733 612-2734
Pipetas volumétricas, BLAUBRAND®, 2 marcas, classe AS
Pipetas Pipetas Volumétricas
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate
DIN EN ISO 648
Certificações: DIN EN ISO 648; DE-M marking.