Print…
Moisture Analyser, MB32
Fornecedor: OHAUS
30971922
611-5848EA 1950 EUR
611-5848
Moisture Analyser, MB32
Analisadores Analisadores de humidade
Perfect for routine moisture testing.
  • An LCD with second line display, navigation knob, and intuitive operation system makes for convenient routine moisture testing
  • The new carbon fiber heating technology and upgraded loadcell shortens analysis time, saves power and keeps results accurate and consistent
  • Robust, slim design with a strong housing ensures MB32's durability even in harsh environments
  • External calibration
The MB32 features a redesigned housing with stronger material, innovative carbon fiber heating technology and an upgraded load cell forMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Capacidade 90 g
Interface de comunicação RS232; USB device
Dimensões 350×210×140 mm
Operating temperature range 10...40 °C
Tamanho do prato 90 mm
Legibilidade 1 mg/0,01%
Repetibilidade 0,02% (10 g sample); 0,15% (3 g sample)
Tensão 240 V