Print…
VWR®, Temporizador/cronómetro digital
Fornecedor: VWR Collection
609-0128EA 27.7 EUR
609-0128
VWR®, Temporizador/cronómetro digital
Temporizadores Temporizadores digitais
Single-channel electronic timer with memory and certificate of calibration.
  • LCD
  • Counts down and up for 99 minutes, 59 seconds
  • Last time set memory
  • Mounting options: Lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment
Resolution: 0,1
Certificações: Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Fornecimento: Supplied withMais informação do produto