Print…
Funnels with level control, b.safe
Fornecedor: Bohlender
R840-15 R840-16 R840-25 R840-26 R880-15 R890-15
590-0381EA 272 EUR
590-0381 590-0382 590-0385 590-0386 590-0403 590-0407
Funnels with level control, b.safe
Funis Funis de segurança
b.safe funnels with level control are made of electrically conductive dissipative PE-EX and suitable for use with flammable liquids. Electrically conductive waste canisters are usually black and therefore the filling volume is not visible. This is where the funnel with fill level control comes in.
  • Prevent the waste canisters from overflowing
  • Available for canisters and barrels
  • With lid or splash protector
  • 130 or 180 mm funnel diameter
The funnels are available in two sizes, each with a hinged lid or splash guard. The unique design with slanted threadMais informação do produto