Print…
Air valves, b.safe
Fornecedor: Bohlender
M505-01 M505-02 M505-05 M505-10
590-0173EA 45 EUR
590-0173 590-0174 590-0175 590-0410
Air valves, b.safe
Válvulas Válvulas de regulação de ar
Valve body made of PP with connection (thread UNF ¼" 28G) suitable for b.safe caps. Lid with aeration opening and Luerlock connector, integrated non-return valve with upstream porous PTFE membrane (1 μm), including clip-on service life indicator.
  • PP for high resistance and fire class rating V-0
  • Non-slip grip area, eroded surfaces and gripping grooves for easy installation
  • Freely movable and individually positionable for all round visibility
  • Activation of service life through touch of the button and/or individual marking on the back side
  • Centring pin for
Mais informação do produto