Print…
Cleanroom wipes, Anticon® MicroQuilt
Fornecedor: CONTEC
495758-737 495758-735 495758-738 495758-736
568-0907EA 1250 EUR
568-0907 568-0905 568-0908 568-0906
Cleanroom wipes, Anticon® MicroQuilt
Toalhetes Toalhetes para salas limpas Seco
Anticon® MicroQuilt combines the cleanliness and sorbent capacity of Quiltec® with the microfibre particle pick-up ability of Anticon® Gold Sorb. This hybrid creates the most sorbent, cleanest wipe Contec offers.
  • Hybrid of polyester and microfibre for the most sorbent and clean wipe Contec offers
  • Laminated 2 ply wipe readily releases fluid to a surface
  • Unique microfibre polyester yarn offers high sorptive capacity and fast absorption rate
  • Sealed edges reduce particle and fibre generation
MicroQuilt is laser-cut with heatsealed edges and cleanroomMais informação do produto