Print…
PEEK frits, Upchurch Scientific®
Fornecedor: UPCHURCH SCIENTIFIC
A-700 A-701 A-702 A-703 A-704 A-706 A-707 A-708 A-710 A-720 A-722 A-735
554-1981EA 24.1 EUR
554-1981 554-1982 UPCHA-702 UPCHA-703 UPCHA-704 554-3049 554-3051 UPCHA-708 UPCHA-710 554-3053 554-3055 UPCHA-735
PEEK frits, Upchurch Scientific®
Filtros Filtros de Pré-Coluna
Exceptionally uniform porosity which ensures longer filtration life and consistent frit-to-frit swept volumes. These frits are biocompatible and inert to most solvents, making them ideal for bioanalytical applications. The disc rings, made of PCTFE, are slightly thicker than the frit disc, providing enhanced sealing and excellent chemical resistance.
  • Inert, biocompatible and metal-free
  • PCTFE surrounded PEEK frits can be used up to 80 °C, others up to 100 °C