Print…
VWR®, Screw Neck Vials ND24
Fornecedor: VWR Collection
548-2357AEA 1170 EUR
548-2357A 548-2428A 548-0156A 548-9000A 548-0638A 548-9002A 548-0639A 548-0640A 548-0637A 548-9001A 548-9003A 548-0641A 548-9004A 548-9005A 548-1462A 548-1463A 548-2532A 548-1057A 548-3235A 548-1090A 548-0825A 548-2145A 548-2142A 548-3237A 548-3236A 548-9025A 548-0154A 548-0155A 548-1390A 548-2452A 548-2536A 548-2537A 548-2538A 548-9024A 548-2400A 548-0634A
VWR®, Screw Neck Vials ND24
Vials
ND24 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
ND24 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom. Associated screw caps, mostly available with assembled septa, are engineered to prevent sample contamination while providing consistent, secure seals.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable