Print…
Extraction thimbles, Munktell
Fornecedor: Ahlstrom-Munksjö
142009
537-4528EA 112 EUR
537-4528
Extraction thimbles, Munktell
Extractores Cartuchos de extracção
Grade ET/401, cellulose.
Particulary suitable for the extraction of organic combinations out of reaction mixtures, food, natural substances, lacquer and binder analysis in paint colour, dust and tar determination in gases.
Max. Temperature: 120 °C
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable