Print…
VWR®, Microcentrifuge Tubes, High Performance
Fornecedor: VWR Collection
525-1161EA 27 EUR
525-1161 525-1162 525-1163 525-1164 525-1165 525-1166 525-1167 525-1168 525-1169 525-1170 525-1171 525-1172 525-1173 525-1174 525-1175 525-1176 525-1177
VWR®, Microcentrifuge Tubes, High Performance
Tubos Tubos de microcentrífuga
These tubes are made from USP Class VI PP, which ensures a clear view of the pellet. They incorporate a high strength wall, and can be centrifuged to twice the g-force that most other microcentrifuge tubes can withstand.
  • Amber coloured for light-sensitive substances
  • Ideal for vortexing and shaking tables
  • RNase-, DNase- and endotoxin-free
  • Engraved graduations ensure accuracy
  • Fit all common rotors
  • Ideal for phenol/chloroform extractions
These tubes also feature flat, pierceable, frosted caps and a side labelling surface. The ergonomically designed caps,Mais informação do produto