Print…
Microcentrifuge tube mini floating racks, Prep-Safe™, SP Bel-Art
Fornecedor: Bel-Art Products, a Part of SP
F187420125
525-1056EA 97.6 EUR
525-1056
Microcentrifuge tube mini floating racks, Prep-Safe™, SP Bel-Art
Suportes/Racks Tube Racks
PP, coloured.
  • PP, steam autoclavable at 121 °C
  • Immobilising tabs prevent tube tops from popping open during boiling
  • Holds up to twelve 1,5 or 2,0 ml microcentrifuge tubes in numbered locations
  • Raised handle for easy lifting: stable feet keep the rack upright on the bench
  • 95 mm outer diameter fits most 1000 ml beakers
Unique tabs protect your samples from loss and contamination. Place tubes in slots and slide under the tabs to keep tubes in place and caps closed when under pressure, floating or submerged. For use in water baths, ice or on the benchtop.
Mais informação do produto