Print…
VWR®, Centrifuge Tubes, Conical Bottom
Fornecedor: VWR Collection
525-0606EA 102 EUR
525-0606 525-0611 525-0607 525-0608 525-0609 525-0610 525-0605 525-0604 525-0612
VWR®, Centrifuge Tubes, Conical Bottom
Tubos Tubos de centrífuga
PP, conical, with white HDPE flat cap or plug cap.
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Caps available in two styles: Flat cap with a smooth surface for additional labelling options, or plug style cap for a secure seal when using shakers and horizontal incubators
  • Engraved graduation at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Can be used for storing and freezing samples at temperatures down to –80 °C
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use
Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable Baixo impacto de fabricação
Certificação ambiental ISO 14001/Certificate No.: 04923E00243R0M