Print…
Vent filters, HEPA-CAP, Whatman™
Fornecedor: Whatman products (Cytiva)
6702-3600 6702-7500 2609T 6702-9500 2709T
516-1104EA 69.75 EUR
516-1104 516-1105 517-0008 517-0012 517-0011
Vent filters, HEPA-CAP, Whatman™
Filtros Filtros de Ventilação
High-retention and high flow-rate glass fiber filters for research and industrial use to remove particulates from air and gases.
  • Typical retention efficiency: 99.97% of all particles ≥ 0.3 µm in air
  • Depth filter design: Allows for high loading capacity and bi-directional flow
  • Repeatedly autoclavable: At 121 °C for 20 minutes (132 °C max.)
Whatman HEPA-CAP Vent Filters are used across scientific, research, and industrial environments in a variety of air and gas filtration applications that require high particle retention and the ability to capture dirtMais informação do produto