Print…
Filtros de seringa Acrodisc®, PTFE
Fornecedor: Cytiva (Formerly Pall Lab)
4219 4225 4226 4422 4423 4472 4501 4503 4521 4542 4543 4552 4553 AP-4219 AP-4302 AP-4303 AP-4501 AP-4518 4555 AP-4225 AP-4521 AP-4791 AP-4790 4219-N 4552TC 4219TC AP-4225TC
PALL4219EA 0 EUR
PALL4219 514-4004 514-4005 514-4008 514-4009 514-4019 516-7651 516-7653 516-7663 516-7671 516-7672 514-4032 514-4033 514-4229 514-4230 514-4231 514-4232 514-0476 514-0086 514-0472 514-0478 514-0186 514-0184 514-4003 514-1439 514-1450 514-1452
Filtros de seringa Acrodisc®, PTFE
Filtros Filtros de Seringa
Exceptional chemical and temperature compatibility. Ideal for filtration of gas and organic solvents. Prevent spurious peaks on chromatograms allowing accurate experimental results. HPLC certified for low level of UV-absorbing extractables.
Hydrophobic PTFE membrane with PP, autoclavable housing
The GxF version provides two to four times the throughput of standard pre-filter devices.
Encomendar
ESPECIFICAÇÕES
Tipo Syringe filter Syringe filter Syringe filter Syringe filter Syringe filter Syringe filter Premium syringe filter (PSF) Premium syringe filter (PSF), with multi-layered glass fibre pre-filter (GxF) Premium syringe filter (PSF) Premium syringe filter (PSF), with multi-layered glass fibre pre-filter (GxF) Premium syringe filter (PSF), with multi-layered glass fibre pre-filter (GxF)
Ø 25 mm 25 mm 25 mm 13 mm 13 mm 4 mm 25 mm 25 mm 25 mm 25 mm 25 mm
Environmentally Preferable Redução de resíduos Redução de resíduos Redução de resíduos Redução de resíduos
Tamanho do poro 0,45 µm 0,2 µm 1,0 µm 0,45 µm 0,2 µm 0,45 µm 0,45 µm 0,45 µm 0,2 µm 0,2 µm 0,2 µm
Max. operating pressure - - - 6,9 bar 6,9 bar 5,2 bar 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C - 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C - 4,1 bar at 21 − 24 °C; 2,1 bar at 100 °C
Max. temperature 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C 100 °C
Taxa de fluxo da água - - - - - - 510 ml/min (1,0 bar) 395 ml/min (1,0 bar) 245 ml/min (1,0 bar) 395 ml/min (1,0 bar) 245 ml/min (1,0 bar)