Print…
Filtros de microfibra de vidro
Fornecedor: Ahlstrom-Munksjö
410122 410121 410124 410123 410037 410052 410054 410056 410057 410058 410015 410005 526016 410073 410008 410067 410204 410013 410134 410157 410003 410095 3.01136.055
513-0750EA 33.4 EUR
513-0750 513-0751 513-0752 513-0754 513-0772 513-0776 513-0777 513-0778 513-0779 513-0780 513-0759 513-0756 513-0781 BINZ410073 BINZ410008 BINZ410067 BINZ410204 BINZ410013 513-0151 BINZ410157 BINZ410003 BINZ410095 BINZ3.01136.055
Filtros de microfibra de vidro
Filtros Filtros de Fibra de Vidro
Ahlstrom-Munksjö offers high quality glass filters ideal for many applications such as air pollution control, water monitoring and air particulate monitoring. The wide range of binder free glass microfiber filters can be used to collect particles and aerosols or to determine total suspended solids in water.
  • Made with ultrapure water
  • Fast flow rate
  • High loading capacity
  • Retention of very fine particles