Print…
General purpose qualitative filter papers
Fornecedor: Ahlstrom-Munksjö
120014 120015 120016 120017 120018 120019 120021 120065 120012 120013 120004 120006 120007 120008 120010 120000 120002 120003 120032 120034 120035 120036 120037 120040 120030 120031 120038 123255 123256 123250 123251 123253 123263 123011 123008 123010 123012 123151 123169 123171 123172 123242 123002 123145 123471 123117 123051 123299 123437 123084 123109 123093 123113 123087 123074 123089 123111 123174 123344 123329 123584 123476 123268 123249 123257 123254 123474 123269
512-6106EA 13.2 EUR
512-6106 512-6105 512-6104 512-6103 512-6102 512-6101 512-6100 512-6109 512-6108 512-6107 512-6096 512-6094 512-6093 512-6092 512-6090 512-6099 512-6098 512-6097 512-6117 512-6114 512-6113 512-6112 512-6111 512-6110 512-6120 512-6119 512-6193 512-6195 512-6196 512-6235 512-6236 512-6238 512-6247 516-0750 516-0748 516-0749 516-0751 516-0757 BINZ123169 BINZ123171 BINZ123172 BINZ123242 BINZ123002 BINZ123145 BINZ123471 BINZ123117 BINZ123051 BINZ123299 BINZ123437 BINZ123084 BINZ123109 BINZ123093 BINZ123113 BINZ123087 BINZ123074 BINZ123089 BINZ123111 BINZ123174 BINZ123344 BINZ123329 BINZ123584 BINZ123476 BINZ123268 516-0767 516-0769 516-0768 BINZ123474 BINZ123269
General purpose qualitative filter papers
Filtros Papéis de Filtro Papel de filtro qualitativo
Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Standard qualitative filterMais informação do produto