Print…
Dispositivos de centrifugação, Vivaspin® 6
Fornecedor: Sartorius
VS0611 VS0621 VS0631 VS0641 VS0612 VS0602 VS0622 VS0632 VS0642 VS0672 VS0601 VS0651 VS0691 VS0692
512-3776EA 250 EUR
512-3776 512-3777 512-3778 512-3779 512-4011 512-4012 512-4013 512-4014 512-4015 512-4017 512-3775 512-4029 512-4103 512-4104
Dispositivos de centrifugação, Vivaspin® 6
Filtros Filtros de Centrifugação
Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in either swing bucket or fixed angle rotors accepting standard 15 ml conical bottom test tubes.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 6 features twin vertical membranes for unparalleled filtration speed and >100-Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Área de filtração 2,5 cm²
Max. RCF 10000 ×g
Volume morto <10 µl
Ø×H 16×122 mm