Print…
Frascos Dewar de transporte, tipo B e B-E
Fornecedor: KGW
1211 1212 1213 1214 1221 1222 1223 1224 1932 1933 1935
478-4301EA 269 EUR
478-4301 478-4302 478-4303 478-4304 478-0765 478-0766 478-0733 478-0767 KGWA1932 KGWA1933 KGWA1935
Frascos Dewar de transporte, tipo B e B-E
Recipientes para armazenamento criogénico
Silvered borosilicate glass, blue coated metal protective covering (B) or stainless steel (B-E). For storage and transport of liquid nitrogen, dry ice and temperature-sensitive goods.
  • Robust design
  • Included insulated lid held by two spring clips
  • With aluminium handle
Nota sobre segurança: Deve ser perfurado um pequeno orifício na tampa, se for armazenado gelo seco ou azoto líquido com a tampa posta.
Safety note: A small hole must be drilled in the lid if dry ice or liquid nitrogen are to be stored with the lid in place.