Print…

Iron, AAS standard solution, Fe 1000 µg/ml, Specpure®

Fornecedor: Thermo Fisher Scientific

47282.AE 47282.AP
47282.AEEA 57.1 EUR
47282.AE 47282.AP
Iron, AAS standard solution, Fe 1000 µg/ml, Specpure®
Padrões Cationic Standards Iron Standards

Molecular Formula: (Fe(NO₃)₃).9H₂O in 5% HNO₃
Physical Form: Liquid
Odour: Acidic
UN No.: UN3265
MDL No.: MFCD00010999

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR