Print…
Drying ovens with natural convection ED series, Avantgarde.Line
Fornecedor: Binder
9010-0333 9010-0335 9010-0339 9010-0341
466-0476EA 1478 EUR
466-0476 466-0477 466-0478 BIND9010-0341
Drying ovens with natural convection ED series, Avantgarde.Line
Estufas Estufas standard
With natural convection, Avantgarde.Line heating and drying chambers offer quality and process reliability. With a wide temperature range up to 300 °C, these drying ovens can be used in all applications. Short heat up times and large power reserves are ideal for applications in science, research and industry. APT.line™ pre-heating chamber technology gives homegeneous temperature distribution and excellent temperature uniformity and fluctuation. Units up to 115 litres can be stacked on top of each other to save space. These units have 60 mm insulation which lowers energy consumption andMais informação do produto