Print…
Banhos e circuladores, aço inoxidável, séries CORIO™ C-B e CD-B
Fornecedor: Julabo
9012419 9011405 9011413 9011417 9011419 9011427 9012405 9012413 9012417 9012427 9012433 9012439
461-0912EA 1898 EUR
461-0912 461-0904 461-0905 461-0906 461-0907 461-0908 461-0909 461-0910 461-0911 461-0913 461-0914 461-0915
Banhos e circuladores, aço inoxidável, séries CORIO™ C-B e CD-B
Termóstatos de circulação Sistemas de circulação aberta Banhos com circuladores amovíveis
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO™ C immersion circulator is the basic model, it is ideal for standard internal applications, the CD model is more advanced and can used for temperature control of external applications if required. The jet nozzle allows continuous adjustment of the pump stream in the bath. The stainless steel baths are durable and have an integrated drain tap (except B5). These open heating bath circulators are ideal for temperature control of samples,Mais informação do produto