Print…

Sulphur in diesel fuel, standard solutions, Specpure®

Fornecedor: Thermo Fisher Scientific

Sulphur in diesel fuel, standard solutions, Specpure®
Padrões Petroleum Industry Standards

Flash Point: 69° (156 °F)
Physical Form: Liquid
Density: 0,6 to 0,9
Appearance: Light brown
Boiling Point: 171 to 315°
MDL No.: MFCD01095233

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR