Print…
Sera-Mag carboxylate-modified magnetic beads and SpeedBeads
Fornecedor: Cytiva
45152105050250 65152105050250 44152105050250 44152105050350 45152105050350 65152105050350 24152105050250 24152105050350
45152105050250.EA 584 EUR
45152105050250. 65152105050250. 44152105050250. 44152105050350. 45152105050350. 65152105050350. 24152105050250. 24152105050350.
Sera-Mag carboxylate-modified magnetic beads and SpeedBeads
Esferas Esferas magnéticas
Sera-Mag Carboxylate-Modified Magnetic Beads & SpeedBeads have free carboxyl groups on the surface for convenient covalent coupling of target molecules (proteins, peptides, and amine-modified ligands) via primary amines (NH2).
  • Carboxylic groups on the surface form covalent amide bonds with proteins, peptides or amine-modified ligands for use in many downstream applications including protein purification, DNA sample preparation and clean-up, protenomicsand immunoassays
  • Carbodiimide chemistry allows rapid formation of stable bonds to minimize ligand