Print…
Rotating mixers, RM 5
Fornecedor: GLASWARENFABRIK KARL HECHT
40348026 40348010 40349018
HECH40348026EA 847 EUR
HECH40348026 445-2510 HECH40349018
Rotating mixers, RM 5
Agitadores Agitadores de balanço/rotativos
Tumbling rotating mixer with five or ten synchronous, diagonally eccentric PVC rolls to provide an uniform and gentle mixing of the tubes.
  • Specially designed for the uniform mixing of medical laboratory tests of blood and haematological determinations and other cell suspension samples
  • Prevents sedimentation in samples such as dyes, paints, oil and cosmetics
Fornecimento: Supplied with cable and safety plug.