Print…
Rocker, Rocker II™
Fornecedor: BOEKEL SCIENTIFIC
260350-EU 260350-UK
444-3512EA 1540 EUR
444-3512 444-3513
Rocker, Rocker II™
Agitadores Agitadores de balanço/rotativos
This is designed for wide variety of flasks, plates and tubes. It is applied for gel staining and de-staining, northern, southern and western blots, mixing test tubes and rocking agglutination card.
  • Adjust tilt angle and speed without the need for tools
  • Low voltage power is supplied by a wall mount transformer
  • Thin power cord is present which is easily passes through the incubator door gaskets
  • Optional stacking tray and dimpled mat (for test tubes) also available
Fornecimento: Supplied with flat non-skid mat.