Print…
VWR® INCU-Line®, Incubadora com agitação
Fornecedor: VWR Collection
444-0732EA 4630 EUR
444-0732 444-0733
VWR® INCU-Line®, Incubadora com agitação
Agitadores Incubadoras com agitação
Shaking incubators with a small footprint. These compact units feature a stackable design and expanded temperature range which makes the ILS 4 and ILS 6 ideal for molecular biology applications and general use. The integral orbital shaker means they are highly versatile. The SmartChek™ temperature control system (exclusive to the Avantor Collection) guarantees precise temperature control. Mechanical convection maintains a stable temperature environment and provides quick recovery after opening the door. A safety thermostat is located on the back of the unit. The insulated door has a large,Mais informação do produto