Print…
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Fornecedor: VWR Collection
391-0637EA 44.8 EUR
391-0637
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Vedantes Selos de microplaca
Easier to apply and remove as compared to standard moulded PCR strip caps.
  • Ideal for PCR
  • User-friendly alternative to strip-caps; no sore thumbs and fingers guaranteed
  • Seals a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Certified DNase- and RNase-free
  • 2,0 mil pierceable foil
  • Recommended temperature range from −20 to +120 °C
6 strips are pre-cut on a continuous liner for ease of use.