Print…
Fernbach culture flasks
Fornecedor: SciLabware
1138/10 1139/10
391-0533EA 482 EUR
391-0533 391-0534
Fernbach culture flasks
Balões Frascos de Cultura
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
  • Exhibit 3 baffle indents to provide optimal and consistent agitation
  • Large surface area to volume ratio
  • Available in two versions, with Delong neck or screw cap