Print…
Frascos de cultura Roux, DURAN®
Fornecedor: DWK Life Sciences
215415801 215715804
391-0360EA 65.9 EUR
391-0360 391-0170
Frascos de cultura Roux, DURAN®
Balões Frascos de Cultura
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Round neck (391-0360)
  • Conical neck, eccentric (391-0170)
Typical application: preparation of cultures in nutrient media.