Print…
Walk-in-chambers for Stability Testing and Storage, WIC
Fornecedor: Binder
9800-0011 9800-0012 9800-0013
390-1702EA 74900 EUR
390-1702 390-1703 390-1704
Walk-in-chambers for Stability Testing and Storage, WIC
Câmaras de testes Câmaras climáticas
The WIC series walk-in-chambers offer a wide range of features and benefits to perfectly meet requirements in stability testing. Perfect test conditions are maintained over a wide range of temperatures from 10 to 50 °C and relative humidity of 20 to 90% RH.
  • Units have relative humidity accuracy of ±2,5% and temperature accuracy of ±1,5 °C
  • Duct work for proper distribution of air
  • Highly insulated room panels with U value of only 0,21 W/min⁻² K
  • LED lighting with motion detector
  • High quality room elements, made in Germany
  • Interior completely stainless steel
Mais informação do produto