Print…

Karl fischer reagents, Fluka™

Fornecedor: Honeywell Chemicals

36115-1L 36116-1L 36116-6X1L 36117-1L
36115-1LEA 140.08 EUR
36115-1L 36116-1L 36116-6X1L 36117-1L
Karl fischer reagents, Fluka™
Titulantes e Reagentes de Karl Fischer

Karl fischer reagents can be used for for pyridine-based volumetric KF titration.


  • Titre: 5,5 to 6,5 mg/ml


Mixture of solutions A and B (ratio 1:1) yield water equivalent (titre) of mi, for pyridine-based volumetric KF titration.

Encomendar

ESPECIFICAÇÕES

Environmentally Preferable
Ponto de ebulição 123 °C >60 °C >60 °C
Ponto de inflamação 33 °C 16 °C 11 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR