Print…

Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornecedor: Tonbo Biosciences
35-8054-U500
35-8054-U500EA 142 EUR
35-8054-U500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Goat anti-Mouse IgG (H&L), F(ab')2 Fragment - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR