Print…
Sampling palette knife spatula, white, single-use, LaboPlast®/SteriPlast®
Fornecedor: BURKLE
5378-1035 5378-0035
331-0368EA 69.8 EUR
331-0368 331-0369
Sampling palette knife spatula, white, single-use, LaboPlast®/SteriPlast®
Espátulas
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle’s volumetric and dosing spoons.
  • PS, white
  • Optionally packaged individually and sterilised
  • L×W 192×20 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
The dimensionally stable long edge of theMais informação do produto