Print…
Sampling scoops, long handle, disposable, LaboPlast®/SteriPlast®
Fornecedor: BURKLE
5378-5014 5378-5013 5378-5018 5378-5017
331-0341EA 31.8 EUR
331-0341 331-0342 331-0343 331-0344
Sampling scoops, long handle, disposable, LaboPlast®/SteriPlast®
Conchas
This sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content, or to reach through narrow openings.
  • PS, white
  • Individually packed and optionally sterilised
  • Handle length: 300 mm
  • For single use
  • Cleanroom manufactured (clean room Class 7)
With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container. The disposable scoop is suitable for sensitive areas of pharmaceuticals and chemistry as well as theMais informação do produto