Print…
Bag closures with scoop/spoon, WeLoc® Scoop
Fornecedor: WELOC
S421100150404 S421500550404 S422201100404 S42320250040
312-0115EA 49.1 EUR
312-0115 312-0116 312-0117 312-0118
Bag closures with scoop/spoon, WeLoc® Scoop
Pinças /Clamps Pinças de uso geral
These sealing clips feature an integrated measuring scoop/spoon. They are convenient for many types of applications in the domestic, medical and industrial field, where both a seal and a measure is required. The clips are made from high quality Polyamide 6.6 material from DuPont® and therefore perfect for use in the demanding pharmaceutical and laboratory environment.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Manufactured in Sweden
Mais informação do produto